Order Entry
Switzerland
ContactUsLinkComponent
Grp78/BiP ELISA kit
Grp78/BiP ELISA kit
Catalog # ENZOADI9002140001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Grp78/BiP ELISA kit
Catalog # ENZOADI9002140001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Grp78/BiP ELISA kit
  • Seq:
    02
  • Sensitivity:
    8,4 ng/ml (1,4-4500 ng/ml)

Specifications

About this item

High-sensitivity ELISA kit for quantifying both normal and upregulated levels of Grp78/BiP for unfolded protein response, cancer and neurodegenerative disease research.

For the quantitative determination of human, mouse and rat Grp78/BiP in cell lysate and serum samples.

Ordering information: Contains microtiter plate, conjugate, antibody, assay buffer, wash buffer concentrate, standard, TMB substrate, stop solution 2, extraction reagent.