ImmunoSet® HSP27 high sensitivity (human), ELISA development set
Catalog # ENZOADI960076
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:1 SET
- Primary antibody reactivity:Human
- Target protein:HSP27
- Description:ImmunoSet® HSP27 high sensitivity (human), ELISA development set
- Seq:02
- Sensitivity:97 pg/ml (range 0,1 - 3,2 ng/ml)
Specifications
About this item
Most comprehensive portfolio of heat shock protein (chaperone) products commercially available.
For the quantitative determination of human HSP27 in cell lysates, plasma, and serum.
The ImmunoSet® HSP27 high sensitivity (human), EIA development set contains sufficient reagents for the development of 5×96 well EIA plates which provides analysis of ~200 samples in duplicate. Save time, money, and sample by developing your own EIA kits. Simply coat your plates and go! This kit provides superior accuracy with fully quantitative results compared to Western blot analysis and has low reactivity with related molecules.
Ordering information: Contains Capture antibody, Standard, Detection antibody, SA-HRP.