PGF2α high sensitivity ELISA kit
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
The PGF2α high sensitivity EIA kit for the quantitative determination of PGF2α in culture supernatants, milk, plasma, serum, saliva, and urine from any species.
- Highly sensitive quantification of PGF2α
- Assay more samples without extraction
The PGF2α high sensitivity EIA kit is a colorimetric competitive enzyme immunoassay kit with results overnight + 3 hours. Absorbance is read at 405 nm.
Ordering information: Kit contains DxS IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution.
Specifications
- Assay Type:Competitive
- Target protein:PGF2α
- Storage conditions:4 °C (long term)
- Sensitivity:0,98 pg/ml (range 1,95 - 2000 pg/ml)
Specifications
Frequently Bought Together