Osteopontin (human), ELISA kit
ENZOADI900142
:
- Pk:1 KIT
- Primary antibody reactivity:Human
- Target protein:Osteopontin
- Description:Osteopontin (human), ELISA kit
- Seq:02
- Sensitivity:0,110 ng/ml (range 2 - 32 ng/mlL)
For the quantitative determination of human Osteopontin in culture supernatants, milk, plasma, and urine. Cited sample type includes serum.
The Osteopontin (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours. Absorbance is read at 450 nm. Ready-to-use liquid color-coded reagents reduce error. Excellent publications in peer-reviewed publications demonstrate the utility and consistency of this kit developed with highly cited double monoclonal antibodies.
: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 10 concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution.