Order Entry
Switzerland
Orders LinkContactUsLinkComponent
TNF alpha (human) ELISA kit

TNF alpha (human) ELISA kit

Supplier:   Enzo Life Sciences
TNF alpha (human) ELISA kit
Supplier:  Enzo Life Sciences
Certificates

2 Options Available Below

Product Details & Documents

Highly sensitive ELISA for measuring TNF-α, the primary mediator of immune regulation.

For the quantitative determination of human TNF-α in culture supernatants, plasma, and serum.

The TNF-α (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 4 hours. Absorbance is read at 450 nm. Assay up to 39 samples in duplicate per 96-well plate. The broad dynamic range makes assaying different pathologies easy.

Ordering information: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 21, Wash buffer concentrate, Standard, TMB Substrate, RIPA Cell lysis buffer 2, Stop solution 2.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • ENZOADI900099
  • ENZOADI901099
  • Target protein:
  • TNF-alpha
  • TNF-alpha
  • Primary antibody reactivity:
  • Human
  • Human
  • Conjugate ELISA:
  • HRP-Streptavidin
  • HRP-Streptavidin
  • Format:
  • Pre-coated
  • Pre-coated
  • Sensitivity:
  • 8,43 pg/ml
  • 8,43 pg/ml
  • Detection range:
  • 15,63 - 1000 pg/ml
  • 15,63 - 1000 pg/ml
  • Detection method:
  • Colorimetric
  • Colorimetric
  • Storage conditions:
  • 4 °C (long term), –20 °C (for standards)
  • 4 °C (long term), –20 °C (for standards)

Specifications

Product Family Options

Product Information

  • Tests per kit
    Pk
    Availability
    Price
  • ENZOADI900099
    TNF-α (human), ELISA kit
    TNF-α (human), ELISA kit
    1×96 wells
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - SFr. 844.00
    SFr. 844.00
    1 * 1 KIT
       SFr. 844.00

    Specifications:
    More
    Specifications
    Cat. No.
    ENZOADI900099
    Target protein
    TNF-alpha
    Primary antibody reactivity
    Human
    Conjugate ELISA
    HRP-Streptavidin
    Format
    Pre-coated
    Sensitivity
    8,43 pg/ml
    Detection range
    15,63 - 1000 pg/ml
    Detection method
    Colorimetric
    Storage conditions
    4 °C (long term), –20 °C (for standards)
  • ENZOADI901099
    TNF-alpha (Human), ELISA kit
    TNF-alpha (Human), ELISA kit
    5×96 wells
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - SFr. 3’440.00
    SFr. 3’440.00
    1 * 1 KIT
       SFr. 3’440.00

    Specifications:
    More
    Specifications
    Cat. No.
    ENZOADI901099
    Target protein
    TNF-alpha
    Primary antibody reactivity
    Human
    Conjugate ELISA
    HRP-Streptavidin
    Format
    Pre-coated
    Sensitivity
    8,43 pg/ml
    Detection range
    15,63 - 1000 pg/ml
    Detection method
    Colorimetric
    Storage conditions
    4 °C (long term), –20 °C (for standards)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.