Order Entry
Switzerland
ContactUsLinkComponent
PGF2α ELISA kit
PGF2α ELISA kit
Catalog # ENZOADI900069
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
PGF2α ELISA kit
Catalog # ENZOADI900069
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Competitive
  • Target protein:
    PGF2α
  • Description:
    PGF2α ELISA kit
  • Seq:
    02
  • Sensitivity:
    6,71 pg/ml (range 3,05 - 50,000 pg/ml)

Specifications

About this item

For the quantitative determination of PGF2α in culture supernatants, plasma, serum, saliva, and urine from any species. Cited sample types include cell lysate and whole blood.

The PGF2α EIA kit is a colorimetric competitive enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 405 nm. Broad dynamic range is suitable for a wide variety of sample types. Ready-to-use liquid color-coded reagents are safe, non-radioactive, and reduce laboratory errors.

Ordering information: Contains DxS IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-316 (5x96 wells)