IL-4 (mouse), ELISA kit
Catalog # ENZOADI900043
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Mouse
- Target protein:IL-4
- Description:IL-4 (mouse), ELISA kit
- Seq:02
- Sensitivity:4,34 pg/ml (range 7,81 - 1000 pg/ml)
Specifications
About this item
For the quantitative determination of mouse IL-4 in culture supernatants and serum. Cited sample type includes plasma.
The IL-4 (mouse), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours.
Ordering information: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 13, Wash buffer concentrate, Standard, TMB Substrate, Stop solution.