Gastrin I (human), ELISA kit
Catalog # ENZOADI900026
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 KIT
- Assay Type:Competitive
- Primary antibody reactivity:Human
- Target protein:Gastrin I
- Description:Gastrin I (human), ELISA kit
- Seq:02
- Sensitivity:7,27pg/ml (range 39,1 - 10000 pg/ml)
Specifications
About this item
For the quantitative determination of human Gastrin I in culture supernatants, plasma, and serum.
The Gastrin I (human), EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 5 hours. Absorbance is read at 405nm. Ready-to-use liquid color-coded reagents are provided to reduce error in the lab.
Ordering information: Contains GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution.