Order Entry
Switzerland
ContactUsLinkComponent
Cysteinyl leukotriene ELISA kit
Cysteinyl leukotriene ELISA kit
Catalog # ENZOADI901070
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Cysteinyl leukotriene ELISA kit
Catalog # ENZOADI901070
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Competitive
  • Description:
    Cysteinyl leukotriene ELISA kit
  • Seq:
    02
  • Sensitivity:
    26,6 pg/ml (range 78,1 - 2,500 pg/ml)

Specifications

About this item

Highly sensitive ELISA kit enabling detection of LTC4/D4/E4 in 4 hours.

For the quantitative determination of Cysteinyl Leukotrienes (LTC4/D4/E4) in culture supernatants and urine from any species.

The Cysteinyl Leukotriene EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 4 hours. Absorbance is read at 405 nm. Safe non-radioactive measurement of LTC4/D4/E4. Ready-to-use liquid color-coded reagents reduce errors.

Ordering information: Contains GxM Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-122 (1x96 wells).