Order Entry
Switzerland
ContactUsLinkComponent
15-deoxy-δ12,14-PGJ2 ELISA kit
15-deoxy-δ12,14-PGJ2 ELISA kit
Catalog # ENZOADI901023
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
15-deoxy-δ12,14-PGJ2 ELISA kit
Catalog # ENZOADI901023
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Competitive
  • Target protein:
    15-deoxy-δ12,14-PGJ2
  • Description:
    15-deoxy-δ12,14-PGJ2 ELISA kit
  • Seq:
    02
  • Sensitivity:
    36,8 pg/ml (range 195 - 200000 pg/ml)

Specifications

About this item

First-to-market 15-d-PGJ2 ELISA commercially available for eicosanoid research.

For the quantitative determination of 15-deoxy-Δ12,14-PGJ2 in culture supernatants, plasma, saliva, and urine from any species.

The 15-deoxy-Δ12,14-PGJ2 EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 5 hours. Absorbance is read at 405 nm. This kit saves time and is much less time consuming than GC/MS analysis. Ready-to-use liquid color-coded reagents reduce laboratory error.

Ordering information: Contains GxR Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-082 (1×96 wells).