NGAL (Human) ELISA kit
Catalog # ENZOBPDKIT036
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Human
- Target protein:NGAL
- Description:NGAL (Human) ELISA kit
- Seq:02
- Storage conditions:4 °C (Long term)
- Sensitivity:4 pg/ml (Range 10 - 1000 pg/ml)
Specifications
About this item
A colorimetric immunometric ELISA kit for the measurement of NGAL in human samples. Absorbance is read at 450 nm.
- Fast results from up to 40 samples in duplicate in just 4 hours per 96 well plate
- Highly sensitive measurement, detecting as low as 4 pg/ml of NGAL
- Ready-to-use pre-coated plate and individual calibrators save time and reduce errors
- Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of NGAL in human urine, plasma, serum, tissue extracts, and culture supernatants.
Delivery: Kit consists of 12 strips of 8 microwells each coated with anti-NGAL (96 wells) + frame, sample diluent, NGAL calibrators 1-8; 0, 10, 25, 50, 100, 250, 500, 1000 pg/ml, 25X wash solution concentrate, biotinylated NGAL antibody, HRP-Streptavidin conjugate, TMB substrate and stop solution.