Order Entry
Switzerland
ContactUsLinkComponent
Corticosterone ELISA kit
Corticosterone ELISA kit
Supplier:  ENZO LIFE SCIENCES
Certificates
undefined
Corticosterone ELISA kit
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Assay Type:
    Competitive
  • Storage conditions:
    4 °C (long term)
  • Sensitivity:
    27,0 pg/ml (range 32 - 20000 pg/ml)

Specifications

Frequently Bought Together