Order Entry
Switzerland
ContactUsLinkComponent
ER-ID™ red assay kit (GFP-CERTIFIED®)
ER-ID™ red assay kit (GFP-CERTIFIED®)
Catalog # ENZOENZ51026K500
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
ER-ID™ red assay kit (GFP-CERTIFIED®)
Catalog # ENZOENZ51026K500
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    ER-ID® Red assay kit (GFP-CERTIFIED®)
  • Seq:
    02
  • Storage conditions:
    –80 °C (Long Term)
  • Application:
    Fluorescence Microscopy
  • Tests per kit:
    500 assays

Specifications

About this item

ER-ID® Red assay kit (GFP-CERTIFIED®) contains an endoplasmic reticulum-selective dye suitable for live cell, or detergent-permeabilized aldehyde-fixed cell staining.

Micromolar concentrations of ER-ID® Red dye are sufficient for staining mammalian cells, as validated with human cervical carcinoma cell line, human T-lymphocyte cell line, Jurkat, HeLa and human bone osteosarcoma epithelial cell line, U2OS. One important application of ER-ID® Red dye is in fluorescence co-localization imaging with green fluorescent protein (GFP)-tagged proteins, a powerful approach for determining the targeting of molecules to intracellular compartments and for screening of their associations and interactions.

The ER-ID® Red assay kit GFP-CERTIFIED®) is specifically designed for use with GFP-expressing cell lines, as well as cells expressing blue, cyan or yellow fluorescent proteins (BFPs, CFPs, YFPs). Additionally, the kit is suitable for use with live or post-fixed cells in conjunction with probes, such as labeled antibodies, or other fluorescent conjugates displaying similar spectral properties as fluorescein, or coumarin. A nuclear counterstain is provided to highlight this organelle as well

Delivery: Kit includes ER-ID® Red detection reagent (50 μl), Hoechst 33342 nuclear stain (50 μl), 10X Assay buffer (15 ml).