Order Entry
Switzerland
ContactUsLinkComponent
ALDetect™ Lipid Peroxidation Assay kit
ALDetect™ Lipid Peroxidation Assay kit
Catalog # ENZOBMLAK1700001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
ALDetect™ Lipid Peroxidation Assay kit
Catalog # ENZOBMLAK1700001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    100 Tests
  • Description:
    PROTEOSTAT® Aggresome detection kit
  • Seq:
    02
  • Storage conditions:
    4 °C (long term)
  • Application:
    Colorimetric detection
  • Size:
    100 tests

Specifications

About this item

The assay is designed to measure either MDA alone (in hydrochloric acid) or MDA in combination with 4-hydroxyalkenals (in methanesulfonic acid). It is based on the reaction of a chromogenic reagent, N-methyl-2-phenylindole with MDA and 4-hydroxyalkenals to yield a stable chromophore with maximal absorbance at 586 nm.

Lipid peroxidation is a well-established mechanism of cellular injury in both plants and animals, and is used as an indicator of oxidative stress in cells and tissues. Lipid peroxides are unstable and decompose to form a complex series of compounds including reactive aldehydes.

Polyunsaturated fatty acid peroxides generate malondialdehyde (MDA) and 4-hydroxyalkenals (HAE) upon decomposition. Measurement of malondialdehyde and 4-hydroxyalkenals has been used as an indicator of lipid peroxidation.

Delivery: Kit includes Reagent R1: N-methyl-2-phenylindole, in acetonitrile (3x18 ml), Reagent R2: conc. methanesulfonic acid (16.5 ml), MDA Standard: 1,1,3,3-tetramethoxypropane (TMOP) in TRIS-hydrochloric acid (1 ml), Diluent: ferric iron in methanol (30 ml).