ApoSENSOR™ Cell viability assay kit
Product Details & Documents
The assay utilizes bioluminescent detection of the ATP level for a rapid screening of apoptosis and cell proliferation simultaneously in mammalian cells. The assay utilizes the enzyme luciferase to catalyze the formation of light from ATP and luciferin. The light can be measured using a luminometer or Beta Counter. The assay can be fully automatic for high throughput (10 seconds/sample) and is highly sensitive (detects 10-100 mammalian cells/well).
Delivery information: Kit includes Nucleotide Releasing Buffer, ATP Monitoring Enzyme, Enzyme Reconstitution Buffer and ATP.
Specifications
Specifications
- Pk:200 Tests
- Description:ApoSENSOR™ cell viability assay kit
- Cat. No.:ENZOALX850247KI01
- Storage conditions:−20 °C (long term)
- Size:200 tests
- Application:Chemiluminescence
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



