Anti-MAVS Mouse Monoclonal Antibody [clone: 58N3B6]
Catalog # ENZOENZABS2590100
Supplier: ENZO LIFE SCIENCES
Specifications
- Antibody type:Primary
- Antigen name:Cardif
- Antigen symbol:MAVS
- Clonality:Monoclonal
- Clone:58N3B6
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human,Mouse
- Western blot:Yes
- Seq:02
- Form:Liquid
- Antigen synonyms:Mitochondrial antiviral signalling protein|Interferon-β promoter stimulator protein 1|VISA|IPS-1|Virus-induced signalling adapter|CARD adapter inducing interferon-β
- Storage buffer:In 0,2 ml PBS containing 0,05% BSA and 0,05% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:Shipped on blue ice
- Immunogen:Human MAVS (IPS-1)
- Purification:Protein G purified
- Pk:100 µG
Specifications
About this item
Recommended Applications: IHC (PS), WB
Type: Primary
Antigen: MAVS
Clonality: Monoclonal
Clone: 58N3B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Mouse