Order Entry
Switzerland
ContactUsLinkComponent
Anti-CD40 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: mAb 89]
Anti-CD40 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: mAb 89]
Catalog # ENZOENZABS1400100
Supplier:  ENZO LIFE SCIENCES
Anti-CD40 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: mAb 89]
Catalog # ENZOENZABS1400100
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CD40
  • Clonality:
    Monoclonal
  • Clone:
    mAb 89
  • Conjugation:
    Alexa Fluor® 647
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Seq:
    02
  • Form:
    Liquid
  • Antigen synonyms:
    Tumor necrosis factor receptor superfamily member 5|B-cell surface antigen CD40|CD40L receptor|TNFRSF 5|Bp50|CDw40
  • Storage buffer:
    In PBS containing 50% glycerol.
  • Storage temperature:
    −80 °C
  • Shipping temperature:
    Shipped on Blue Ice
  • Immunogen:
    Anti-μ activated tonsillar human B cells.
  • Purification:
    Purified using QMA Hyper D ion exchange chromatography
  • Size:
    200 µl
  • Pk:
    100 µG

Specifications

About this item

Recommended Applications: Flow Cytometry, IF

Species reactivity: Human

Type: Primary
Antigen: CD40
Clonality: Monoclonal
Clone: mAb 89
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: