Order Entry
Switzerland
ContactUsLinkComponent
Anti-GHRL Mouse Monoclonal Antibody [clone: 45]
Anti-GHRL Mouse Monoclonal Antibody [clone: 45]
  ENZOBPDABS050451
Anti-GHRL Mouse Monoclonal Antibody [clone: 45]
  ENZOBPDABS050451

 

  • Antibody type:
    Primary
  • Antigen name:
    Ghrelin
  • Antigen symbol:
    GHRL
  • Clonality:
    Monoclonal
  • Clone:
    45
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 lambda
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Seq:
    03
  • Form:
    Liquid
  • Antigen synonyms:
    ghrelin 28|obestatin|ghrelin 27|Growth hormone-releasing peptide|MTLRP|Protein M46|Motilin-related peptide|Appetite-regulating hormone
  • Storage buffer:
    0,01 M phosphate buffer, pH 7,4, containing 0,5 M sodium chloride and 15 mM sodium azide
  • Storage temperature:
    4 °C
  • Shipping temperature:
    Blue ice not frozen
  • Immunogen:
    Synthetic peptide corresponding to the sequence of human human Ser3-octanoyl ghrelin-28
  • Purification:
    Purified by Protein A affinity chromatography
  • Pk:
    1 mg

 

 

Type: Primary
Antigen: Ghrelin
Clonality: Monoclonal
Clone: 45
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human