- Antibody type:Primary
- Antigen name:Lipocalin 2
- Antigen symbol:LCN2
- Clonality:Monoclonal
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Rat
- Western blot:Yes
- Seq:03
- Form:Liquid, in 0,01 M phosphate buffer, pH 7,4, containing 0,5 M sodium chloride and 15 mM sodium azide
- Antigen synonyms:lipocalin 2|oncogenic lipocalin 24p3|migration-stimulating factor inhibitor|MSFI|siderocalin|uterocalin|neutrophil gelatinase-associated lipocalin|lipocalin 2 (oncogene 24p3)|LCN2|oncogene 24p3|NGAL
- Purification:Protein A/G-affinity purified
- Pk:1 mg
Rat lipocalin-2 is also called rat NGAL, as it is the ortholog of human neutrophil gelatinase-associated lipocalin (NGAL). Rat NGAL is a 25-kDa alpha 2-microglobulin-related protein or neu-related lipocalin. It is expressed by neutrophils and various epithelial cells, including cells of the renal proximal tubules. It is involved in physiological processes such as the binding of siderophore iron and tissue growth and regeneration. It is upregulated in a variety of pathological situations including infection, inflammation, ischemia-reperfusion and in certain adenocarcinomas.
- For ELISA, Flow Cytometry, Western Blot
Recognizes rat NGAL. Detects a band of ~25kDa by Western blot.
Type: Primary
Antigen: LCN2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat