Order Entry
Switzerland
ContactUsLinkComponent
Anti-CDH5 Mouse Monoclonal Antibody
Anti-CDH5 Mouse Monoclonal Antibody
Codice catalogo ENZOALX803305C100
Nome del fornitore:  ENZO LIFE SCIENCES
Anti-CDH5 Mouse Monoclonal Antibody
Codice catalogo ENZOALX803305C100
Nome del fornitore:  ENZO LIFE SCIENCES

Specifiche

  • Antibody type:
    Primary
  • Antigen name:
    Cadherin 5
  • Antigen symbol:
    CDH5
  • Clonality:
    Monoclonal
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid. In PBS containing 1 mg/ml BSA and 0,02% sodium azide.
  • Antigen synonyms:
    Cd144 Antigen|Endothelial-Specific Cadherin|Vascular Endothelial Cadherin|VE-Cadherin|CD144
  • Concentration:
    0,1 mg/ml
  • Purification:
    Affinity purified
  • Pk:
    100 µG

Specifiche

Informazioni su questo articolo

Host: Mouse, Isotype: IgG2a

  • For ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human VE-cadherin. Does not cross-react with other species. Detects a band of ~130-140kDa by Western blot.

Type: Primary
Antigen: CDH5
Clonality: Monoclonal
Clone: 0
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: