Restriction enzymes, SmaI, FastDigest™, Fermentas
Catalog # FD0663
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 KIT
- Application:
- Compatible buffer:FastDigest Green Buffer or FastDigest Buffer (colorless)
- Overhang type:Blunt
- Source:
- Description:SmaI, FastDigest™
- Environmentally Preferable:
- Enzyme name:SmaI
- No. of reactions:100 (100 µl)
Specifications
About this item
Thermo Scientific™ FastDigest™ enzymes are an advanced line of restriction enzymes for rapid DNA digestion.
- 100% activity of all FastDigest™ enzymes in one universal reaction buffer
- Direct loading on gels with FastDigest™ Green buffer
- 100% buffer compatibility with downstream applications
Designed to complete digestion in 5 to 15 minutes.