Restriction enzymes, SalI, FastDigest™, Fermentas
FD0644
:
- Pk:1 KIT
- Application:
- Compatible buffer:FastDigest Green Buffer or FastDigest Buffer (colorless)
- Overhang type:5' overhang
- Source:
- Description:SalI, FastDigest™
- Environmentally Preferable:
- Enzyme name:SalI
- No. of reactions:200 (200 µl)
Thermo Scientific™ FastDigest™ enzymes are an advanced line of restriction enzymes for rapid DNA digestion.
- 100% activity of all FastDigest™ enzymes in one universal reaction buffer
- Direct loading on gels with FastDigest™ Green buffer
- 100% buffer compatibility with downstream applications
Designed to complete digestion in 5 to 15 minutes.