Order Entry
Switzerland
ContactUsLinkComponent
 
Benzethonium chloride 0.01 N, AVS TITRINORM volumetric solution
Catalog # 89576.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
Benzethonium chloride 0.01 N, AVS TITRINORM volumetric solution
Catalog # 89576.320
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Titer:
    0.01000 ±0.5%

Specification Test Results

About this item

 :
 :448.09 g/mol
CAS Number:121-54-0
Formula:C₂₇H₄₂ClNO₂
MDL Number:MFCD00011742