Order Entry
Switzerland
ContactUsLinkComponent
Magnesium chloride, anhydrous, white crystalline powder
Magnesium chloride, anhydrous, white crystalline powder
Catalog # ICNA0520984480
Supplier:  MP Biomedicals
CAS Number:  
Magnesium chloride, anhydrous, white crystalline powder
Catalog # ICNA0520984480
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Appearence:
    White anhydrous crystalline powder
  • Solubility:
    Soluble in exothermic, water (4 g/10 ml-clear colourless haze free solution)

Specification Test Results

About this item

 :
 :95.21 g/mol
Storage Temperature:Ambient
CAS Number:7786-30-3
Formula:MgCl₂
MDL Number:MFCD00011106