ContactUsLinkComponent
VWR®, Filter Holder, Stainless Steel
VWR®, Filter Holder, Stainless Steel
Catalog # 511-0666
Supplier:  VWR Collection
undefined
VWR®, Filter Holder, Stainless Steel
Catalog # 511-0666
Supplier:  VWR Collection
Specifications
  • Description:
    Glass filter holder with stainless steel screen
  • Pk:
    1
Specifications
About this item
Glass filter holder with 47 mm stainless steel screen, 300 ml glass funnel, 47 mm aluminium spring clamp, 47 mm glass base, PTFE gasket and silicone stopper no. 8.