ContactUsLinkComponent
Papanicolaou's solution 2a Orange G (OG 6), Sigma-Aldrich®
Flammable
Irritant
Papanicolaou's solution 2a Orange G (OG 6), Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Flammable
Irritant
Papanicolaou's solution 2a Orange G (OG 6), Sigma-Aldrich®
Supplier:  Merck
Certificates
Specification Test Results
  • Suitability for microscopy (vaginal smear):
    Passes test
  • Nuclei:
    Blue to dark violet
  • Cyanophilic cytoplasm:
    Blue-green
  • Eosinophilic cytoplasm:
    Pink