Order Entry
Switzerland
ContactUsLinkComponent
Arsenic standard solution, for limit tests
Arsenic standard solution, for limit tests
Codice catalogo REGE5000500C
Nome del fornitore:  REAGECON
Numero CAS:  
Arsenic standard solution, for limit tests
Codice catalogo REGE5000500C
Nome del fornitore:  REAGECON
Numero CAS:  

Specifiche

  • Concentration:
    10 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    100 mL
  • Element:
    Arsenic
  • Symbol:
    As

Specifiche

Informazioni su questo articolo

Concentrate to make arsenic standard solution (10 ppm as As) as outlined in Chapter 4 (4.1.2) of Ph. Eur.