Order Entry
Switzerland
ContactUsLinkComponent
Potassium iodate 0.05 M in aqueous solution
Potassium iodate 0.05 M in aqueous solution
Catalog # REGEPI2005F
Supplier:  REAGECON
CAS Number:  
Potassium iodate 0.05 M in aqueous solution
Catalog # REGEPI2005F
Supplier:  REAGECON
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Molarity:
    0,05 M

Specification Test Results

About this item

 :
 :214 g/mol
CAS Number:7758-05-6
Formula:KlO₃
MDL Number:MFCD00011406