Order Entry
Switzerland
ContactUsLinkComponent
Reticulum stain kit acc. to Gordon & Sweets, Sigma-Aldrich®
Health
Corrosive
Irritant
Environment
Reticulum stain kit acc. to Gordon & Sweets, Sigma-Aldrich®
Catalog # 1.00251.0001
Supplier:  Merck
CAS Number:  
undefined
Health
Corrosive
Irritant
Environment
Reticulum stain kit acc. to Gordon & Sweets, Sigma-Aldrich®
Catalog # 1.00251.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Fibre case
  • Pk:
    24 Tests
  • Seq:
    0765

Specifications

Specification Test Results

  • Suitability for microscopy:
    Passes test
  • Reticular fibres:
    Black
  • Nuclei:
    Red
  • Collagen:
    Red
  • Background:
    Red

Specification Test Results

About this item

 :
UN:3316
ADR:9,II
 :GHS08
 :GHS05
 :GHS07
 :GHS09
 :1