ContactUsLinkComponent
Biotin picolyl azide
Biotin picolyl azide
Catalog # 92186.
Supplier:  Biotium
CAS Number:  
Biotin picolyl azide
Catalog # 92186.
Supplier:  Biotium
CAS Number:  
Specifications
  • Pk:
    0,5 mg
  • Description:
    CF™ Biotin picolyl azide
  • Seq:
    10
Specifications
About this item
Biotin picolyl azide reacts with alkyne to form 1,2,3-triazole by 1,3-dipolar Huisgen cycloaddition through the use of a much lower copper (I) concentration without sacrificing reaction efficiency.