ContactUsLinkComponent
Dehydrated culture media, R-2A agar suitable for microbiology, NutriSelect® Plus
Dehydrated culture media, R-2A agar suitable for microbiology, NutriSelect® Plus
Catalog # SIAL17209-500G
Supplier:  Merck
CAS Number:  
Dehydrated culture media, R-2A agar suitable for microbiology, NutriSelect® Plus
Catalog # SIAL17209-500G
Supplier:  Merck
CAS Number:  
Specifications
  • Pk:
    500 g
  • Description:
    R-2A agar
  • Water:
    +
Specifications
About this item
R-2A agar is used for heterotrophic plate count of water samples using longer incubation periods.