
Sodium tetrathionate dihydrate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13721-29-4
:
Formula:Na₂O₆S₄·2H₂O
MDL Number:MFCD00150868
:306.27 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Solubility:(1 g in 20 ml water) Clear to hazy
- Titration after Ion exchange:≥97.5 %
Specification Test Results
Frequently Bought Together