Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

7-Hydroxyindole 95%

Corrosive
Irritant
7-Hydroxyindole 95%
Certificates
7-Hydroxyindole 95%

2 Options Available Below

Product Details & Documents
  • CAS Number:2380-84-9
  • Formula:C₈H₇NO
  • MDL Number:MFCD00152102
  • Molecular Weight:133.15 g/mol
  • Melting Pt:85…87 °C
  • Storage Temperature:Freezer
Specifications for Product Family

Specification Test Results

  • Appearance:
    Yellow to brown Solid or crystalline solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    83 to 88 °C
  • GC:
    ≥94.0 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - SFr. 141.00
    SFr. 141.00
    1 * 250 mg
       SFr. 141.00

  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - SFr. 422.00
    SFr. 422.00
    1 * 1 g
       SFr. 422.00