Lids, Silicone Gaskets, and Screw Caps for Standard Cells
Supplier: HELLMA
New Product
Certificates
About this item
An assortment of lids, silicone gaskets, and screw caps designed for use with standard Hellma® cells.
Hellma® precision cells are 100% manufactured in Germany and are suitable for all common spectrophotometers
- Hellma® precision cells are characterized by the highest precision in path length accuracy and parallelism
- Hellma® precision cells can be easily cleaned and reused and thus remain in use for a long time
Open screw caps are suitable for use with cell types 117.100, 117.100F, 117.104, 117.104F.
Closed screw caps are suitable for use with cell types 117.200, 117.200F, 117.204, 117.204F.
Replacement gaskets are suitable for use with cell types 117.100, 117.100F, 117.104, 117.104F, 117.200, 117.200F, 117.204, 117.204F.
Frequently Bought Together