Screw caps for culture tubes, KIMBLE®
Catalog # SCERSP73805B-15415
Supplier: DWK Life Sciences
Specifications
- Thread:15-415
- Colour:Natural
- Description:With closed top, linerless
- Material:PP
- Pk:1.000
Specifications
About this item
PP.
- Very good chemical resistance
- Very good temperature resistance