Tri-camp GL-stopcocks, BOLA
Supplier: Bohlender
Certificates
About this item
Made of PTFE/PP.
- Very good chemical resistance
- FDA confirmity
- Temperature resistant from 0 to +110 °C
- Quick and easy disconnection of flow
Cylindrical 2-Way stopcock with straight bore, Tri-Clamp and GL-threaded neck, made of PTFE for reliable tightness and grip made of PP for marking the flow direction.
Certifications: DIN 32 676
Frequently Bought Together