Storage vials, round, Azlon®
Supplier: SciLabware
Certificates
About this item
These are highly transparent PC vials with coloured PP cap. Ideal for sampling, production and packaging of steam sterilised biological solutions.
- Leakproof and shatterproof
- Suitable for repeated sterilisation
- Withstands temperatures to –80 °C
Delivery: Supplied with non sterile plastic wrapped trays with PP caps.
Frequently Bought Together