ContactUsLinkComponent
Cleanroom wipes, C3
Cleanroom wipes, C3
Catalog # EPTA7-C3-99L-00
undefined
Cleanroom wipes, C3
Catalog # EPTA7-C3-99L-00
Specifications
  • Colour:
    White
  • L×W:
    229×229 mm
  • Pk:
    2.400
Specifications
About this item
These 100% hydroentangled polyester wipes offer the highest level of softness and abrasion resistance − perfect for cleaning and polishing the most sensitive surfaces.
  • Tough and absorbent for usage in a variety of applications
  • Recommended for the cleaning of lenses or other sensitive surfaces
  • Chemically compatible with most cleaning solutions
  • Low NVRs and ions
  • No chemical binders
Typical applications include: Optics cleaning and manufacturing, LCD and display cleaning and maintenance, sensitive surface wipe downs and prep, use as a work surface for delicate tasks, critical tool and component wipe downs.
Recommended for use in ISO 5 - 8 controlled environments.