ContactUsLinkComponent
Reagents for fluoride determination using Lovibond® MD photometers
Reagents for fluoride determination using Lovibond® MD photometers
Catalog # LOVI467481
Supplier:  Lovibond
CAS Number:  
undefined
Reagents for fluoride determination using Lovibond® MD photometers
Catalog # LOVI467481
Supplier:  Lovibond
CAS Number:  
Specifications
  • Pk:
    250 mL
  • Description:
    SPADNS reagent solution
  • Seq:
    44
  • For unit:
    MD 100, MD 600, MD610, MultiDirect and SpectroDirect
  • Range:
    0,2 – 2 mg/l F