ContactUsLinkComponent
Ammonium molybdate reagent for silica determination using Lovibond₂ Comparator 2000+ system
Ammonium molybdate reagent for silica determination using Lovibond₂ Comparator 2000+ system
Catalog # LOVI460241
Supplier:  Lovibond
CAS Number:  
undefined
Ammonium molybdate reagent for silica determination using Lovibond₂ Comparator 2000+ system
Catalog # LOVI460241
Supplier:  Lovibond
CAS Number:  
Specifications
  • Chemical method:
    Silicomolybdate
  • Pk:
    100 mL
  • Description:
    Silica reagent, ammonium molybate liquid solution
  • Range:
    2.5 – 25 mg/l SiO₂