ContactUsLinkComponent
Reagents for dissolved oxygen determination using Lovibond® MD photometers
Reagents for dissolved oxygen determination using Lovibond® MD photometers
Catalog # LOVI380450
Supplier:  Lovibond
CAS Number:  
undefined
Reagents for dissolved oxygen determination using Lovibond® MD photometers
Catalog # LOVI380450
Supplier:  Lovibond
CAS Number:  
Specifications
  • Chemical method:
    Rhodazine D
  • Pk:
    30 Tests
  • Description:
    Dissolved oxygen Vacu-vial® K-7553 test kit
  • Environmentally Preferable:
  • For unit:
    MD600, MD610, MD640 and MultiDirect
  • Range:
    10 – 800 µg/l O2