Reagents for urea determination using Lovibond® MD photometers,
Supplier: Lovibond
Certificates
About this item
Frequently Bought Together
Neutralising agent for the automated cleaning of glassware, neodisher® N
Supplier: Dr. Weigert
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
pH indicator, phenol red tablets and solutions, Lovibond®
Supplier: Lovibond
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Stem Thermometer, Calibrated, Electronic, Stainless Steel, 127 mm Probe
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Powder packs for determination of chlorine, reagents for photometers
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
Sodium thiosulphate, anhydrous 99%
Supplier: Thermo Fisher Scientific
o-Cresolphthalein complexone tetrasodium salt, pure
Supplier: Thermo Fisher Scientific
FerroZine Powder, 500 g 1 * 500 g
Supplier: Hach
N,N-Diethyl-p-phenylenediammonium sulphate 97%
Supplier: Thermo Fisher Scientific
Single channel macro pipettes, fixed / variable volume, Acura® manual 835F and 835
Supplier: Socorex Isba
S-Phenyl-L-cysteine 97%
Supplier: Thermo Fisher Scientific