Reagents for DEHA determination with Lovibond® MD photometers
Supplier: Lovibond
Certificates
Certificates
About this item
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® High Performance, Bottles, Wide Neck with Screw Cap
Supplier: VWR Collection
Test kits, iron, Spectroquant®, Supelco®
Supplier: Merck
Test strips for glucose, MQuant®, Supelco®
Supplier: Merck
VWR®, Chloride 1,000 mg/L in Water standard for ion chromatography
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Syringes, 3-piece
Supplier: TERUMO
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Test strips, sulphite, MQuant®, Supelco®
Supplier: Merck
VWR®, Sulphate 1000 mg/L in water standard for ion chromatography
Supplier: VWR Chemicals
Sodium acetate ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Pipette Tips, Standard
Supplier: VWR Collection
Colour calibration standard solutions, Pt-Co, Supelco®
Supplier: Merck
Test kits, phosphate (kits and reagents) CHECKIT®
Supplier: Lovibond
Accessories for water purification millipore
Supplier: Merck
Iodine 0.05 M I2 (0.1N) volumetric solution, Fluka™
Supplier: Honeywell Chemicals