Shoe covers, 442
Catalog # MMMA442
Supplier: 3M
Specifications
- Anti-static:+
- Garment application:General Purpose,Medical,Laboratory,Industrial,Food
- Colour:White
- Height:180 mm
- Length:380 mm
- Pk:300
- Fabric:PP/PE
- Weight:55 g/m²
Specifications
About this item
These disposable shoe covers are designed to provide basic barrier protection to help protect footwear against light liquid splashes and hazardous dusts. Made from a microporous laminate material (PP/PE) with anti-static treatment.
- Elastic around the ankle for protection against contaminants and enhanced fit
- Sole and upper constructed of the same material
- Anti-static treated
Weight: 55 g/m²
Length: 380 mm
Height: 180 mm
Opening diameter: 8 - 15 cm