Vydac 214MS C4, HPLC columns
Supplier: Avantor
Certificates
About this item
These Vydac MS columns are a later development from Grace for the reversed phase HPLC separation of biomolecules. A proprietary surface treatment and bonding process give Vydac MS columns unique selectivity. A variety of reversed phases makes this product line suitable for the analysis of small peptides to large intact, undenatured proteins.
- Five reversed-phase chemistries
- Excellent peak shape with little or no TFA
- High protein recoveries make scale-up easy
Lower capacity than C18 or C8, suitable for hydrophobic proteins or when minimal organic solvent is desired. Ideal for biomolecules >10K MW, undenatured intact proteins, antibodies, oligonucleotides, human growth hormone.
Vydac is a registered trademark of W.R.Grace and Co.
Specifications
- Catalog No:
- 214MS5405
- 214MS5.315
- 214MS5415
- 214MS5.1505
- 214MS1022
- 214MS5215
- 214GD54MS/N
- 214GK52MS/N
- 214GK54MS/N
- 214MS5.07515
- 214MS5205
- 214GD52MS/N
- 214FSK1210MS
- 214MS5.325
- 214MS54
- 214MS52
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Carbon load:
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- USP code:
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
Specifications
Frequently Bought Together