Vydac 219TP Diphenyl, HPLC columns
Supplier: Avantor
Certificates
About this item
Vydac TP was the first range of wide pore media phases developed in the Vydac range and became the industry standard for peptide and protein separations. The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.
- Long column lifetime and negligible phase leaching
- Reliable protein purifications, scalable from analytical to preparative scale
- Referenced in a large number of patents and publications
Lowest capacity first generation diphenyl phase. Recommended for polypeptides with aromatic side chains, large hydrophobic proteins, membrane-spanning peptides, lipid peptides, fusion proteins from inclusion bodies.
Vydac is a registered trademark of W.R.Grace and Co.
Specifications
- Catalog No:
- 219TP54
- 219TP510
- 219GD52/N
- 219GCC510
- 219GK52/N
- 219TP5215
- 219TP5210
- 219TP1010
- 219TP5415
- 219FSK1210
- Carbon load:
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- USP code:
- L11
- L11
- L11
- L11
- L11
- L11