ContactUsLinkComponent
Avantor® Genesis, HPLC Columns, AQ
Avantor® Genesis, HPLC Columns, AQ
Supplier:  Avantor
   Sustainable Options Available
Certificates
undefined
Avantor® Genesis, HPLC Columns, AQ
Supplier:  Avantor
   Sustainable Options Available
Specifications
  • Catalog No:
  • FM15951E
  • FH1951-2/N
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Carbon load:
  • 15%
  • Endcapped:
  • Yes
  • pH range:
  • 1 - 10
  • Surface area:
  • 300 m²/g
  • Environmental certification:
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)