Vacuum seal PTFE / FKM
Codice catalogo 531-9423
Nome del fornitore: Heidolph Instruments GmbH & Co.KG
Specifiche
- Description:Standard spare vacuum seal, FKM
- Environmentally Preferable:
- Pk:1
Specifiche
Informazioni su questo articolo
The highly durable and long lasting PTFE vacuum seals with FKM sealing ring achieve maximum tightness and save on long-term spare parts costs. The combination of both materials allows for very good chemical resistance, while at the same time offering the best sealing properties.
- Standard PTFE spare seal (with FKM sealing ring)
- Particularly high chemical resistance
- Long running time without abrasion
- Highest density