Anion exchange columns, Dionex™, IonPac™ AS14A
Supplier: DIONEX
Certificates
About this item
IonPac™ AS14A carbon eluent deliver fast isocratic separation of inorganic anions from diverse and complex matrices such as drinking water, wastewater, foods and beverages, and scrubber solutions.
- Separation of common inorganic anions in 8 minutes (3×150 mm format) or 13 minutes (4×250 mm format) in diverse sample matrices
- Superior retention and quantification of fluoride
- Compatible with sodium or potassium hydroxide gradient to optimize difficult separations
- Solvent compatibility permits easy column cleanup after the analysis of samples with hydrophobic components
Specifications
- Catalog No:
- DION56904
- DION56897
- DION56901
- Max. pressure:
- 4000 psi
- 4000 psi
- 4000 psi
- pH range:
- 2 – 11
- 2 – 11
- 2 – 11
Specifications
Frequently Bought Together