Single channel pipettes, mechanical, variable / fixed volume, Reference® 2 (General Lab Product)
About this item
The robust Reference® 2 is the successor to the Reference® pipette and features a new design, reduced weight and unique one-button operation.
- Single button operation and spring loaded tip cone for fast and ergonomic handling with reduced operating effort
- Quick tip ejection with active aerosol reduction; haptic feedback prevents accidential tip ejection
- Serial number is printed on multiple components of the pipette for heightened traceability
- Embedded RFID chip containing all relevant data ensures complete traceability
- Optional TrackIT software allows simple identification and documentation and provides ability to add own data (e.g. calibration time and results, location)
- Autoclavable
Due to its reliable and accurate operation, the pipette is ideal for use with precious liquids or in any application requiring extremely high levels of accuracy.
Designed and constructed for general laboratory and research applications in Life Science. Typical applications include: Food and beverages, forensic, cosmetics, agriculture, animal studies, chemical analytics, research applications at medical and academic facilities.
Works with windows 7/8/vista.
Certifications: Without CE marking.
Ordering information: All adjustable volume pipettes come with an epT.I.P.S.® box (except the 2,5 ml, 5 ml and 10 ml versions).
Each 3-pack contains three pipettes, one set of matching eppendorf tips and one ballpoint pen.
32 Options Available Below
Product Details & Documents
The robust Reference® 2 is the successor to the Reference® pipette and features a new design, reduced weight and unique one-button operation.
- Single button operation and spring loaded tip cone for fast and ergonomic handling with reduced operating effort
- Quick tip ejection with active aerosol reduction; haptic feedback prevents accidential tip ejection
- Serial number is printed on multiple components of the pipette for heightened traceability
- Embedded RFID chip containing all relevant data ensures complete traceability
- Optional TrackIT software allows simple identification and documentation and provides ability to add own data (e.g. calibration time and results, location)
- Autoclavable
Due to its reliable and accurate operation, the pipette is ideal for use with precious liquids or in any application requiring extremely high levels of accuracy.
Designed and constructed for general laboratory and research applications in Life Science. Typical applications include: Food and beverages, forensic, cosmetics, agriculture, animal studies, chemical analytics, research applications at medical and academic facilities.
Works with windows 7/8/vista.
Certifications: Without CE marking.
Ordering information: All adjustable volume pipettes come with an epT.I.P.S.® box (except the 2,5 ml, 5 ml and 10 ml versions).
Each 3-pack contains three pipettes, one set of matching eppendorf tips and one ballpoint pen.
Documents
Specifications for Product Family
Specifications
- Catalog No:
- 613-5832
- EPPE4924000.908
- 613-5833
- EPPE4924000.916
- 613-5834
- EPPE4924000.924
- 613-5835
- 613-5836
- 613-5837
- 613-5838
- 613-5839
- 613-5840
- 613-5841
- 613-5842
- EPPE4925000.014
- EPPE4925000.022
- EPPE4925000.030
- EPPE4925000.049
- EPPE4925000.057
- EPPE4925000.065
- EPPE4925000.073
- EPPE4925000.081
- EPPE4925000.090
- EPPE4925000.103
- EPPE4925000.111
- EPPE4925000.120
- EPPE4925000.138
- EPPE4925000.146
- EPPE4925000.154
- EPPE4925000.162
- EPPE4925000.170
- 613-7086
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
Specifications
Product Family Options
Variable volume
Product Information
- VolumeAccuracyImprecisionColour codeRecommended tipsAvailabilityPrice
- 0,1 - 2,5 µl0,1 µl: ±48 % / 0,048 µl
0,25 µl: ±12 % / 0,03 µl
1,25 µl: ±2,5 % / 0,031 µl
2,5 µl: ±1,4 % / 0,035 µl0,1 µl: ±12 % / 0,012 µl
0,25 µl: ±6,0 % / 0,015 µl
1,25 µl: ±1,5 % / 0,019 µl
2,5 µl: ±0,7 % / 0,018 µlDark greyepT.I.P.S.® 10 µlLog in for availability1 * 1 STSFr. 414.00Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-5832Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - 0,5 - 10 µl0,5 µl: ±8,0 % / 0,04 µl
1 µl: ±2,5 % / 0,025 µl
5 µl: ±1,5 % / 0,075 µl
10 µl: ±1,0 % / 0,1 µl0,5 µl: ±5,0 % / 0,025 µl
1 µl: ±1,8 % / 0,018 µl
5 µl: ±0,8 % / 0,004 µl
10 µl: ±0,4 % / 0,04 µlGreyepT.I.P.S.® 20 µlLog in for availability1 * 1 STSFr. 414.00Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-5833Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - 2 - 20 µl2 µl: ±3,0 % / 0,06 µl
10 µl: ±1,0 % / 0,10 µl
20 µl: ±0,8 % / 0,16 µl2 µl: ±1,5 % / 0,03 µl
10 µl: ±0,6 % / 0,06 µl
20 µl: ±0,3 % / 0,06 µlLight greyepT.I.P.S.® 20 µlLog in for availability1 * 1 STSFr. 414.00Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-5834Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - 2 - 20 µl2 µl: ±5,0 % / 0,1 µl
10 µl: ±1,2 % / 0,12 µl
20 µl: ±1,0 % / 0,2 µl2 µl: ±1,5 % / 0,03 µl
10 µl: ±0,6 % / 0,06 µl
20 µl: ±0,3 % / 0,06 µlYellowepT.I.P.S.® 200 µlLog in for availability1 * 1 STSFr. 414.00Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-5835Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - 10 - 100 µl10 µl: ±3,0 % / 0,3 µl
50 µl: ±1,0 % / 0,5 µl
100 µl: ±0,8 % / 0,8 µl10 µl: ±1,0 % / 0,1 µl
50 µl: ±0,3 % / 0,15 µl
100 µl: ±0,2 % / 0,2 µlYellowepT.I.P.S.® 200 µlLog in for availability1 * 1 STSFr. 414.00Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-5836Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - 20 - 200 µl20 µl: ±2,5 % / 0,5 µl
100 µl: ±1,0 % / 1,0 µl
200 µl: ±0,6 % / 1,2 µl20 µl: ±0,7 % / 0,14 µl
100 µl: ±0,3 % / 0,3 µl
200 µl: ±0,2 % / 0,4 µlYellowepT.I.P.S.® 200 µlLog in for availability1 * 1 STSFr. 414.00Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-5837Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - 30 - 300 µl30 µl: ±2,5 % / 0,75 µl
150 µl: ±1,0 % / 1,5 µl
300 µl: ±0,6 % / 1,8 µl30 µl: ±0,7 % / 0,21 µl
150 µl: ±0,3 % / 0,45 µl
300 µl: ±0,2 % / 0,6 µlOrangeepT.I.P.S.® 300 µlLog in for availability1 * 1 STSFr. 414.00Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-5838Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - 100 - 1000 µl100 µl: ±3,0 % / 3,0 µl
500 µl: ±1,0 % / 5,0 µl
1000 µl: ±0,6 % / 6,0 µl100 µl: ±0,6 % / 0,6 µl
500 µl: ±0,2 % / 1,0 µl
1000 µl: ±0,2 % / 2,0 µlBlueepT.I.P.S.® 1000 µlLog in for availability1 * 1 STSFr. 414.00Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-5839Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - 250 - 2500 µl250 µl: ±4,8 % / 12 µl
1250 µl: ±0,8 % / 10 µl
2500 µl: ±0,6 % / 15 µl250 µl: ±1,2 % / 3 µl
1250 µl: ±0,2 % / 2,5 µl
2500 µl: ±0,2 % / 5 µlRedepT.I.P.S.® 2500 µlLog in for availability1 * 1 STSFr. 414.00Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-5840Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - 500 - 5000500 µl: ±2,4 % / 12 µl
2500 µl: ±1,2 % / 30 µl
5000 µl: ±0,6 % / 30 µl500 µl: ±0,6 % / 3 µl
2500 µl: ±0,25 % / 6,25 µl
5000 µl: ±0,15 % / 7,5 µlPurpleepT.I.P.S.® 5000 µlLog in for availability1 * 1 STSFr. 414.00Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-5841Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - 1000 - 10000 µl1000 µl: ±3,0 % / 30 µl
5000 µl: ±0,8 % / 40 µl
10000 µl: ±0,6 % / 60 µl1000 µl: ±0,6 % / 6 µl
5000 µl: ±0,2 % / 10 µl
10000 µl: ±0,15 % / 15 µlTurquoiseepT.I.P.S.® 10000 µlLog in for availability1 * 1 STSFr. 414.00Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-5842Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact
3-packs
Product Information
- VolumeAccuracyImprecisionColour codeRecommended tipsAvailabilityPrice
- EPPE4924000.9083-pack, option 1, incl. 3 Eppendorf Reference® 2 pipettes (0,5 - 10 µl, 10 - 100 µl, 100 - 1000 µl), epT.I.P.S.® boxes and ballpoint pen3-pack, option 1, incl. 3 Eppendorf Reference® 2 pipettes (0,5 - 10 µl, 10 - 100 µl, 100 - 1000 µl), epT.I.P.S.® boxes and ballpoint penLog in for availability1 * 1 KITSFr. 970.00Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.EPPE4924000.908Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - EPPE4924000.9163-pack, option 2, incl. 3 Eppendorf Reference® 2 pipettes (2 - 20 µl, 20 - 200 µl, 100 - 1000 µl), epT.I.P.S.® boxes and ballpoint pen3-pack, option 2, incl. 3 Eppendorf Reference® 2 pipettes (2 - 20 µl, 20 - 200 µl, 100 - 1000 µl), epT.I.P.S.® boxes and ballpoint penLog in for availability1 * 1 KITSFr. 970.00Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.EPPE4924000.916Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - EPPE4924000.9243-pack, option 3, incl. 3 Eppendorf Reference® 2 pipettes (100 - 1000 µl, 0,5 - 5 ml, 1 - 10 ml), epT.I.P.S.® samples and ballpoint pen3-pack, option 3, incl. 3 Eppendorf Reference® 2 pipettes (100 - 1000 µl, 0,5 - 5 ml, 1 - 10 ml), epT.I.P.S.® samples and ballpoint penLog in for availability1 * 1 KITSFr. 970.00Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.EPPE4924000.924Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact
Fixed volume
Product Information
- VolumeAccuracyImprecisionColour codeRecommended tipsAvailabilityPrice
- Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.EPPE4925000.014Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.EPPE4925000.022Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.EPPE4925000.030Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.EPPE4925000.049Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.EPPE4925000.057Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.EPPE4925000.065Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.EPPE4925000.073Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.EPPE4925000.081Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.EPPE4925000.090Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.EPPE4925000.103Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.EPPE4925000.111Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.EPPE4925000.120Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.EPPE4925000.138Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.EPPE4925000.146Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.EPPE4925000.154Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.EPPE4925000.162Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.EPPE4925000.170Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact
6-packs
Product Information
- VolumeAccuracyImprecisionColour codeRecommended tipsAvailabilityPrice
- 613-7086Eppendorf Reference® 2, 6-pack with pipette carousel 2, 1-channel, variable, including 6× epT.I.P.S.® box 2.0 and ballpoint pen, 0,1 - 2,5 µl, 0,5 - 10 µl, 2 - 20 µl, 10 - 100 µl, 20 - 200 µl, 100 - 1000 µlEppendorf Reference® 2, 6-pack with pipette carousel 2, 1-channel, variable, including 6× epT.I.P.S.® box 2.0 and ballpoint pen, 0,1 - 2,5 µl, 0,5 - 10 µl, 2 - 20 µl, 10 - 100 µl, 20 - 200 µl, 100 - 1000 µlYellow-Log in for availability1 * 1 KITSFr. 2’120.00Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.613-7086Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRenewable Energy Certificate, ISO 14001Low Manufacturing Impact
Accessories
Product Information
- Catalog #AvailabilityPrice
- Protection filter for 0,25 - 2,5 ml variable / 2 ml fixed volume / 2,5 ml fixed volume, colour code red, 1 set = 10 filters, 1 sleeveProtection filter for 0,25 - 2,5 ml variable / 2 ml fixed volume / 2,5 ml fixed volume, colour code red, 1 set = 10 filters, 1 sleeveEPPE4920622.009Log in for availability1 * 1 ST - SFr. 18.70
Product Information
- Catalog #AvailabilityPrice
- Lower part, 1 - 10 ml variable, turquoiseLower part, 1 - 10 ml variable, turquoiseEPPE4920611.007Log in for availability1 * 1 ST - SFr. 68.50
Product Information
- Catalog #AvailabilityPrice
- Protection filter for 0,5 - 5 ml variable, colour code purple, 1 set = 10 filters, 1 sleeveProtection filter for 0,5 - 5 ml variable, colour code purple, 1 set = 10 filters, 1 sleeveEPPE4920623.005Log in for availability1 * 1 ST - SFr. 18.70
Product Information
- Catalog #AvailabilityPrice
- Pipette holder for pipette carousel
Product Information
- Catalog #AvailabilityPrice
- Protection filter for 1 - 10 ml variable, colour code turquoise, 1 set = 10 filters, 1 sleeveProtection filter for 1 - 10 ml variable, colour code turquoise, 1 set = 10 filters, 1 sleeveEPPE4920624.001Log in for availability1 * 1 ST - SFr. 18.70
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..